Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3′,3′′,5′,5′′-Tetraiodophenolphthalein 95.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
386-17-4
C20H10I4O4
821.914
MFCD00001614
FWQKRBDABCRWKV-UHFFFAOYSA-N
Iodophene, Iodophthalein, Tetraiodophthalein
67846
3,3-bis(4-hydroxy-3,5-diiodophenyl)-2-benzofuran-1-one
C1=CC=C2C(=C1)C(=O)OC2(C3=CC(=C(C(=C3)I)O)I)C4=CC(=C(C(=C4)I)O)I
386-17-4
C20H10I4O4
821.914
MFCD00001614
FWQKRBDABCRWKV-UHFFFAOYSA-N
Iodophene, Iodophthalein, Tetraiodophthalein
67846
3,3-bis(4-hydroxy-3,5-diiodophenyl)-2-benzofuran-1-one
C1=CC=C2C(=C1)C(=O)OC2(C3=CC(=C(C(=C3)I)O)I)C4=CC(=C(C(=C4)I)O)I
Color Yellow
Quantity 25g
Formula Weight 821.91
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (HPLC,T)
Chemical Name or Material 3′,3′′,5′,5′′-Tetraiodophenolphthalein

RTECSNumber : SM8397000

TSCA : No