Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3′,3′′,5′,5′′-Tetraiodophenolsulfonphthalein 80.0+%, TCI America™

Click to view available options
Quantity:
1g
10g
4430-24-4
C19H10I4O5S
857.962
MFCD00010405
RXNYILISXGUYFG-UHFFFAOYSA-N
iodophenol blue,3',3,5',5-tetraiodophenolsulfonephthalein,3,3-bis 4-hydroxy-3,5-diiodophenyl benzo c 1,2-oxathiolene-1,1-dione,phenol, 4,4'-3h-2,1-benzoxathiol-3-ylidene bis 2,6-diiodo-, s,s-dioxide,tetraiodophenolsulfonaphthalein,4,4'-1,1-dioxido-3h-2,1-benzoxathiole-3,3-diyl bis 2,6-diiodophenol,3,3',5,5'-tetraiodophenolsulfonephthalein,3,3',5,5',-tetraiodophenolsulphonephthalein,4-3-4-hydroxy-3,5-diiodophenyl-1,1-dioxo-2,1
78158
4-[3-(4-hydroxy-3,5-diiodophenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2,6-diiodophenol
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)I)O)I)C4=CC(=C(C(=C4)I)O)I
4430-24-4
C19H10I4O5S
857.962
MFCD00010405
RXNYILISXGUYFG-UHFFFAOYSA-N
iodophenol blue,3',3,5',5-tetraiodophenolsulfonephthalein,3,3-bis 4-hydroxy-3,5-diiodophenyl benzo c 1,2-oxathiolene-1,1-dione,phenol, 4,4'-3h-2,1-benzoxathiol-3-ylidene bis 2,6-diiodo-, s,s-dioxide,tetraiodophenolsulfonaphthalein,4,4'-1,1-dioxido-3h-2,1-benzoxathiole-3,3-diyl bis 2,6-diiodophenol,3,3',5,5'-tetraiodophenolsulfonephthalein,3,3',5,5',-tetraiodophenolsulphonephthalein,4-3-4-hydroxy-3,5-diiodophenyl-1,1-dioxo-2,1
78158
4-[3-(4-hydroxy-3,5-diiodophenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2,6-diiodophenol
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)I)O)I)C4=CC(=C(C(=C4)I)O)I
Quantity 10g
Color Undesignated
Melting Point 229°C
Formula Weight 857.96
Physical Form Crystal-Powder at 20°C
Percent Purity ≥80.0% (HPLC)
Chemical Name or Material 3′,3′′,5′,5′′-Tetraiodophenolsulfonphthalein