Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3′,3′′,5′,5′′-Tetraiodophenolsulfonphthalein 80.0+%, TCI America™
Click to view available options
Quantity:
1g
10g
Specifications
Quantity | 1g |
Color | Undesignated |
Melting Point | 229°C |
Formula Weight | 857.96 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥80.0% (HPLC) |
Chemical Name or Material | 3′,3′′,5′,5′′-Tetraiodophenolsulfonphthalein |
Safety and Handling
TSCA : No