Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(3,5-Difluorophenyl)propionic acid, 97%

Manufacturer:  Alfa Aesar™ H5400414

Quantity 25g

 View more versions of this product

Catalog No. AAH5400414


  Check Availability
Add to cart

Specifications

Description

Applications
3-(3,5-Difluorophenyl)propionic acid is used as intermediate in pharmaceuticals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

84315-24-2
186.16
3-(3,5-Difluorophenyl)propionic acid
C9H8F2O2
MFCD04116054

Specifications

60°C
84315-24-2
186.16
3-(3,5-Difluorophenyl)propionic acid
25g
C9H8F2O2
MFCD04116054
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H318-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit