Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5-Triiodo-L-thyronine, sodium salt, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
55-06-1
C15H11I3NNaO4
672.959
MFCD00002594
SBXXSUDPJJJJLC-YDALLXLXSA-M
liothyronine sodium,cytomel,triostat,tertroxin,3,3',5-triiodo-l-thyronine sodium salt,basoprocin,ibiothyron,cynomel,cytomel sodium,triiodothyronine sodium
23666110
CHEBI:6484
sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+]
55-06-1
C15H11I3NNaO4
672.959
MFCD00002594
SBXXSUDPJJJJLC-YDALLXLXSA-M
liothyronine sodium,cytomel,triostat,tertroxin,3,3',5-triiodo-l-thyronine sodium salt,basoprocin,ibiothyron,cynomel,cytomel sodium,triiodothyronine sodium
23666110
CHEBI:6484
sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+]
Specific Rotation Condition +20.00 (20.00°C c=2,alcohol/1.2N HCl)
Packaging Glass bottle
Melting Point 205.0°C
Loss on Drying 4% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Color White to Beige
Quantity 1g
Specific Rotation + 20.00
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Merck Index 15, 5565
Formula Weight 672.96
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 3,3',5-Triiodo-L-thyronine, sodium salt
Show More Show Less