Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3′,5-Triiodothyroacetic Acid, Approx. 95%, Spectrum™

Click to view available options
Quantity:
100 mg
500 mg
1 g
Packaging:
Amber Glass Bottle
51-24-1
C14H8I3NaO4
643.92
BBBAIWCYEJXUMH-UHFFFAOYSA-M
sodium 2-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetate
[Na+].OC1=CC=C(OC2=C(I)C=C(CC([O-])=O)C=C2I)C=C1I
51-24-1
C14H8I3NaO4
643.92
BBBAIWCYEJXUMH-UHFFFAOYSA-M
sodium 2-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetate
[Na+].OC1=CC=C(OC2=C(I)C=C(CC([O-])=O)C=C2I)C=C1I

T2778, 51-24-1, C14H9I3O4

Spectrum™ 3,3′,5-Triiodothyroacetic Acid
Packaging Amber Glass Bottle
Quantity 1 g
CAS Max % 1
Percent Purity Approx. 95%