Alfa Aesar™ 3-[3-(Benzyloxy)phenyl]propionic acid, 96%

Manufacturer: Alfa Aesar™ H33899MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33899MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57668-34-5
    Molecular Formula C16H16O3
    Formula Weight 256.3
    MDL Number MFCD09258905
    Chemical Name or Material 3-[3-(Benzyloxy)phenyl]propionic acid

    Specifications

    Formula Weight 256.3
    Molecular Formula C16H16O3
    MDL Number MFCD09258905
    CAS 57668-34-5
    Melting Point 83° to 85°C
    Quantity 250mg
    Chemical Name or Material 3-[3-(Benzyloxy)phenyl]propionic acid
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit