Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(chloromethyl)phenyl]-1-methyl-1H-pyrazole, Tech., Maybridge™

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
912569-61-0
C11H11ClN2
206.673
MFCD09702402
XEIKNZPKJWESHW-UHFFFAOYSA-N
3-3-chloromethyl phenyl-1-methyl-1h-pyrazole,3-3-chloromethyl phenyl-1-methylpyrazole
24229632
3-[3-(chloromethyl)phenyl]-1-methylpyrazole
CN1C=CC(=N1)C2=CC(=CC=C2)CCl
912569-61-0
C11H11ClN2
206.673
MFCD09702402
XEIKNZPKJWESHW-UHFFFAOYSA-N
3-3-chloromethyl phenyl-1-methyl-1h-pyrazole,3-3-chloromethyl phenyl-1-methylpyrazole
24229632
3-[3-(chloromethyl)phenyl]-1-methylpyrazole
CN1C=CC(=N1)C2=CC(=CC=C2)CCl
Quantity 250mg
Packaging Amber Glass Bottle
CAS Min % 80.0
CAS Max % 100.0
Formula Weight 206.67
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-[3-(chloromethyl)phenyl]-1-methyl-1H-pyrazole

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger