Alfa Aesar™ 3,3'-Di-n-heptyloxacarbocyanine iodide, 97%

Manufacturer: Alfa Aesar™ H55777ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH55777ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53213-83-5
    Formula Weight 600.58
    MDL Number MFCD00274413
    Chemical Name or Material 3,3'-Di-n-heptyloxacarbocyanine iodide

    Specifications

    Formula Weight 600.58
    Quantity 500mg
    Sensitivity Light Sensitive
    Physical Form Solid
    Color Red
    MDL Number MFCD00274413
    CAS 53213-83-5
    Chemical Name or Material 3,3'-Di-n-heptyloxacarbocyanine iodide
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit