Alfa Aesar™ 3,3'-Diethylthiadicarbocyanine iodide, 99%

As a solvent soluble laser dye

Manufacturer: Alfa Aesar™ 4385104

Quantity 2g

 View more versions of this product

Catalog No. AA4385104

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 514-73-8
    Molecular Formula C23H23N2S2I
    Formula Weight 518.47
    MDL Number MFCD00074829
    UN Number UN2811
    Beilstein 3838938
    Merck Index 14,3369
    Synonym 3-Ethyl-2-[5-[3-ethyl-3H-benzothiazol-2-ylidene]-1,3-pentadienyl]benzothiazolium iodide
    Chemical Name or Material 3,3'-Diethylthiadicarbocyanine iodide

    Specifications

    Formula Weight 518.47
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Molecular Formula C23H23N2S2I
    CAS 514-73-8
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    MDL Number MFCD00074829
    Beilstein 3838938
    Color Green
    Merck Index 14,3369
    Health Hazard 1 Danger
    Solubility Information Soluble in methanol
    Health Hazard 2 GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 2g
    TSCA TSCA
    UN Number UN2811
    Melting Point 248°C (dec.)
    Synonym 3-Ethyl-2-[5-[3-ethyl-3H-benzothiazol-2-ylidene]-1,3-pentadienyl]benzothiazolium iodide
    RTECS Number DL7060000
    Chemical Name or Material 3,3'-Diethylthiadicarbocyanine iodide
    Assay 99%

    GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit