Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3'-Diethylthiadicarbocyanine perchlorate, 96%

Manufacturer:  Alfa Aesar™ H3242103

Quantity 1g

 View more versions of this product

Catalog No. AAH3242103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

22268-65-1
491.12
3,3'-Diethylthiadicarbocyanine perchlorate
C23H23ClN2O4S2
MFCD06409265

Specifications

1g
C23H23ClN2O4S2
MFCD06409265
96%
22268-65-1
491.12
3,3'-Diethylthiadicarbocyanine perchlorate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit