Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,3'-Diethylthiadicarbocyanine perchlorate, 96%, Thermo Scientific™
Click to view available options
Quantity:
250mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 250mg |
Formula Weight | 491.12 |
Percent Purity | 96% |
Chemical Name or Material | 3,3'-Diethylthiadicarbocyanine perchlorate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures