Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dimethoxybenzidine dihydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
20325-40-0
C14H16N2O2·2HCl
317.21
MFCD00012488
UXTIAFYTYOEQHV-UHFFFAOYSA-N
3,3'-dimethoxybenzidine dihydrochloride,o-dianisidine dihydrochloride,3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-qv96qa6ukk,dianisidine dihydrochloride,ccris 998,o-dianisidine hcl,qv96qa6ukk,c.i. disperse black 6 dihydrochloride
62311
4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl
20325-40-0
C14H16N2O2·2HCl
317.21
MFCD00012488
UXTIAFYTYOEQHV-UHFFFAOYSA-N
3,3'-dimethoxybenzidine dihydrochloride,o-dianisidine dihydrochloride,3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-qv96qa6ukk,dianisidine dihydrochloride,ccris 998,o-dianisidine hcl,qv96qa6ukk,c.i. disperse black 6 dihydrochloride
62311
4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl
Melting Point 268.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Color Beige to Gray-Purple
Quantity 5g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 13, 807
Formula Weight 317.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3, 3'-Dimethoxybenzidine dihydrochloride, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 243-737-5

RTECSNumber : DD1050000

TSCA : TSCA