Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-butanol 95.0+%, TCI America™
SDP

Catalog No. p-7126683
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 624-95-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002928
InChI Key DUXCSEISVMREAX-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096
PubChem CID 12233
IUPAC Name 3,3-dimethylbutan-1-ol
SMILES CC(C)(C)CCO

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 mL
UN Number 2282
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butanol
EINECSNumber (2)-0217
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.