Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
5g
887144-97-0
C10H10F3IO
330.09
HVAPLSNCVYXFDQ-UHFFFAOYSA-N
3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole,1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole,togni's reagent,togni reagent,unii-3ok0e02mt6,1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl,1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole,3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole,3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole,togni's reagents dimethyl compound mi
16043572
3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C
887144-97-0
C10H10F3IO
330.09
HVAPLSNCVYXFDQ-UHFFFAOYSA-N
3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole,1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole,togni's reagent,togni reagent,unii-3ok0e02mt6,1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl,1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole,3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole,3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole,togni's reagents dimethyl compound mi
16043572
3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C
Infrared Spectrum Authentic
Quantity 250mg
Melting Point 75.0°C to 79.0°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 330.09
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning