Alfa Aesar™ 3,3-Dimethyl-2-butanol, 99%

Manufacturer: Alfa Aesar™ 4372509

Quantity 10g

 View more versions of this product

Catalog No. AA4372509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 464-07-3
    Molecular Formula (CH3)3CCH(OH)CH3
    Formula Weight 102.18
    MDL Number MFCD00004522
    UN Number UN1987
    Synonym tert-Butyl methyl carbinol; Pinacolyl alcohol
    Chemical Name or Material 3,3-Dimethyl-2-butanol

    Specifications

    Formula Weight 102.18
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    RTECS Number EL2276000
    MDL Number MFCD00004522
    CAS 464-07-3
    UN Number UN1987
    Health Hazard 1 Warning
    Flash Point 28°C (84°F)
    Boiling Point 119° to 121°C
    TSCA TSCA
    Physical Form Liquid
    Molecular Formula (CH3)3CCH(OH)CH3
    Refractive Index 1.4150
    Density 0.812g/mL
    Melting Point 5.3°C
    Synonym tert-Butyl methyl carbinol; Pinacolyl alcohol
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 10g
    Chemical Name or Material 3,3-Dimethyl-2-butanol
    Assay 99%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit