Alfa Aesar™ (+/-)-3,3-Dimethyl-2-butylamine, 98%

Manufacturer: Alfa Aesar™ A1153014

Quantity 25g

 View more versions of this product

Catalog No. AAA1153014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3850-30-4
    Molecular Formula C6H15N
    Formula Weight 101.19
    MDL Number MFCD00008078
    UN Number UN2733
    Synonym 2-Amino-3,3-dimethylbutane
    Chemical Name or Material (±)-3,3-Dimethyl-2-butylamine

    Specifications

    Formula Weight 101.19
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 3850-30-4
    Molecular Formula C6H15N
    MDL Number MFCD00008078
    Quantity 25g
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    TSCA TSCA
    UN Number UN2733
    Synonym 2-Amino-3,3-dimethylbutane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material (±)-3,3-Dimethyl-2-butylamine
    Assay 98%

    Description

    Applications
    (+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

    Solubility
    Fully miscible in water.

    Notes
    Air Sensitive. Store away from acid chlorides and acid anhydrides.

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit