Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylbutylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
15673-00-4
C6H15N
101.19
MFCD00008201
GPWHFPWZAPOYNO-UHFFFAOYSA-N
3,3-dimethylbutylamine,1-butanamine, 3,3-dimethyl,3,3-dimethyl butylamine,3,3-dimethyl-butylamine,3,3-dimethylbutyamine,3,3 dimethylbutylamine,acmc-1btoo,3,3-dimethylbutyl amine,1-butanamine,3,3-dimethyl
19709
3,3-dimethylbutan-1-amine
CC(C)(C)CCN
15673-00-4
C6H15N
101.19
MFCD00008201
GPWHFPWZAPOYNO-UHFFFAOYSA-N
3,3-dimethylbutylamine,1-butanamine, 3,3-dimethyl,3,3-dimethyl butylamine,3,3-dimethyl-butylamine,3,3-dimethylbutyamine,3,3 dimethylbutylamine,acmc-1btoo,3,3-dimethylbutyl amine,1-butanamine,3,3-dimethyl
19709
3,3-dimethylbutan-1-amine
CC(C)(C)CCN
Quantity 5g
Specific Gravity 0.752
Boiling Point 114°C to 116°C
Flash Point 6°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4120 to 1.4160 (20°C, 589nm)
Density 0.7520g/mL
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula (CH3)3CCH2CH2NH2
Formula Weight 101.19
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3, 3-Dimethylbutylamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 239-749-5