Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dimethyldiphenylamine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
626-13-1
C14H15N
197.281
MFCD00059315
CWVPIIWMONJVGG-UHFFFAOYSA-N
di-m-tolylamine,3,3'-dimethyldiphenylamine,m,m'-ditolylamine,dim-tolylamine,bis 3-methylphenyl amine,3-methyl-n-3-methylphenyl aniline,benzenamine, 3-methyl-n-3-methylphenyl,di-m-tolyl-amine,3,3'-ditolylamine,pubchem8963
7016139
3-methyl-N-(3-methylphenyl)aniline
CC1=CC(=CC=C1)NC2=CC=CC(=C2)C
626-13-1
C14H15N
197.281
MFCD00059315
CWVPIIWMONJVGG-UHFFFAOYSA-N
di-m-tolylamine,3,3'-dimethyldiphenylamine,m,m'-ditolylamine,dim-tolylamine,bis 3-methylphenyl amine,3-methyl-n-3-methylphenyl aniline,benzenamine, 3-methyl-n-3-methylphenyl,di-m-tolyl-amine,3,3'-ditolylamine,pubchem8963
7016139
3-methyl-N-(3-methylphenyl)aniline
CC1=CC(=CC=C1)NC2=CC=CC(=C2)C
Refractive Index 1.6210 to 1.6230 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.0400g/mL
Color Yellow
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4NHC6H4CH3
Formula Weight 197.28
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3, 3';-Dimethyldiphenylamine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.