Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylglutaric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
4839-46-7
C7H12O4
160.17
MFCD00002716
DUHQIGLHYXLKAE-UHFFFAOYSA-N
3,3-dimethylglutaric acid,pentanedioic acid, 3,3-dimethyl,3,3-dimethylglutarate,3,3-dimethyl-pentanedioic acid,.beta.,.beta.-dimethylglutaric acid,3,3-dimethyl-glutaric acid,3,3-dimethylpentaneodioic acid,glutaric acid,3-dimethyl,pentanedioic acid,3-dimethyl,beta,beta-dimethylglutaric acid
20984
CHEBI:68502
3,3-dimethylpentanedioic acid
CC(C)(CC(=O)O)CC(=O)O
4839-46-7
C7H12O4
160.17
MFCD00002716
DUHQIGLHYXLKAE-UHFFFAOYSA-N
3,3-dimethylglutaric acid,pentanedioic acid, 3,3-dimethyl,3,3-dimethylglutarate,3,3-dimethyl-pentanedioic acid,.beta.,.beta.-dimethylglutaric acid,3,3-dimethyl-glutaric acid,3,3-dimethylpentaneodioic acid,glutaric acid,3-dimethyl,pentanedioic acid,3-dimethyl,beta,beta-dimethylglutaric acid
20984
CHEBI:68502
3,3-dimethylpentanedioic acid
CC(C)(CC(=O)O)CC(=O)O
Color White to Beige
Boiling Point 290.0°C
Water 1% max. (K.F.)
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Melting Point 98.0°C to 102.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HO2CCH2C(CH3)2CH2CO2H
Beilstein 02, 684
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions
Formula Weight 160.17
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3, 3-Dimethylglutaric acid
Show More Show Less