Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3-Dimethylglutaric anhydride, 97%

Click to view available options
Quantity:
5g
25g
100g
4160-82-1
C7H10O3
142.154
MFCD00006684
HIJQFTSZBHDYKW-UHFFFAOYSA-N
3,3-dimethylglutaric anhydride,2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl,4,4-dimethyldihydro-2h-pyran-2,6 3h-dione,glutaric anhydride, 3,3-dimethyl,3,3-dimethylglutaricanhydride,glutaric anhydride, beta,beta-dimethyl,acmc-1adve,dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione,3,3-dimethyladipic anhydride,3,3 dimethylglutaric anhydride
77813
4,4-dimethyloxane-2,6-dione
CC1(CC(=O)OC(=O)C1)C
4160-82-1
C7H10O3
142.154
MFCD00006684
HIJQFTSZBHDYKW-UHFFFAOYSA-N
3,3-dimethylglutaric anhydride,2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl,4,4-dimethyldihydro-2h-pyran-2,6 3h-dione,glutaric anhydride, 3,3-dimethyl,3,3-dimethylglutaricanhydride,glutaric anhydride, beta,beta-dimethyl,acmc-1adve,dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione,3,3-dimethyladipic anhydride,3,3 dimethylglutaric anhydride
77813
4,4-dimethyloxane-2,6-dione
CC1(CC(=O)OC(=O)C1)C
Boiling Point 181°C (25mmHg)
Quantity 5g
Melting Point 122°C to 126°C
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylglutaric anhydride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA