Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylglutaric Anhydride 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4160-82-1
C7H10O3
142.154
MFCD00006684
HIJQFTSZBHDYKW-UHFFFAOYSA-N
3,3-dimethylglutaric anhydride,2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl,4,4-dimethyldihydro-2h-pyran-2,6 3h-dione,glutaric anhydride, 3,3-dimethyl,3,3-dimethylglutaricanhydride,glutaric anhydride, beta,beta-dimethyl,acmc-1adve,dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione,3,3-dimethyladipic anhydride,3,3 dimethylglutaric anhydride
77813
4,4-dimethyloxane-2,6-dione
CC1(CC(=O)OC(=O)C1)C
4160-82-1
C7H10O3
142.154
MFCD00006684
HIJQFTSZBHDYKW-UHFFFAOYSA-N
3,3-dimethylglutaric anhydride,2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl,4,4-dimethyldihydro-2h-pyran-2,6 3h-dione,glutaric anhydride, 3,3-dimethyl,3,3-dimethylglutaricanhydride,glutaric anhydride, beta,beta-dimethyl,acmc-1adve,dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione,3,3-dimethyladipic anhydride,3,3 dimethylglutaric anhydride
77813
4,4-dimethyloxane-2,6-dione
CC1(CC(=O)OC(=O)C1)C
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 25g
Melting Point 122°C to 126°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 17, 419
Formula Weight 142.15
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 3, 3-Dimethylglutaric anhydride, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 223-998-1

TSCA : TSCA