Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3-Dimethylglutarimide, 99%

Click to view available options
Quantity:
5g
25g
1123-40-6
C7H11NO2
141.17
MFCD00006671
YUJCWMGBRDBPDL-UHFFFAOYSA-N
3,3-dimethylglutarimide,2,6-piperidinedione, 4,4-dimethyl,glutarimide, 3,3-dimethyl,4,4-dimethyl-2,6-piperidinedione,beta,beta-dimethylglutarimide,3,3-dimethyl-glutarimide,.beta.,.beta.-dimethylglutarimide,4,6-piperidinedione,2, 4,4-dimethyl,glutarimide,3-dimethyl
14292
4,4-dimethylpiperidine-2,6-dione
CC1(CC(=O)NC(=O)C1)C
1123-40-6
C7H11NO2
141.17
MFCD00006671
YUJCWMGBRDBPDL-UHFFFAOYSA-N
3,3-dimethylglutarimide,2,6-piperidinedione, 4,4-dimethyl,glutarimide, 3,3-dimethyl,4,4-dimethyl-2,6-piperidinedione,beta,beta-dimethylglutarimide,3,3-dimethyl-glutarimide,.beta.,.beta.-dimethylglutarimide,4,6-piperidinedione,2, 4,4-dimethyl,glutarimide,3-dimethyl
14292
4,4-dimethylpiperidine-2,6-dione
CC1(CC(=O)NC(=O)C1)C
Applications
3,3′-dimethylglutarimide were used as the barbital subsitutes. 3,3-di­methyl­glutarimide is involved formation of the heterodimer.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Melting Point 144°C to 146°C
Quantity 25g
Beilstein 111981
Formula Weight 141.17
Percent Purity 99%
Chemical Name or Material 3,3-Dimethylglutarimide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MA4348000