Alfa Aesar™ 3,3-Dimethylglutarimide, 99%

Manufacturer: Alfa Aesar™ B2219606

Quantity 5g

 View more versions of this product

Catalog No. AAB2219606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1123-40-6
    Molecular Formula C7H11NO2
    Formula Weight 141.17
    MDL Number MFCD00006671
    Beilstein 111981
    Synonym 4,4-Dimethylpiperidine-2,6-dione
    Chemical Name or Material 3,3-Dimethylglutarimide

    Specifications

    Formula Weight 141.17
    CAS 1123-40-6
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number MA4348000
    Quantity 5g
    Molecular Formula C7H11NO2
    MDL Number MFCD00006671
    Beilstein 111981
    Synonym 4,4-Dimethylpiperidine-2,6-dione
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 144° to 146°C
    Chemical Name or Material 3,3-Dimethylglutarimide
    Assay 99%

    Description

    Applications
    3,3 ft.-dimethylglutarimide were used as the barbital subsitutes. 3,3-di­methyl­glutarimide is involved formation of the heterodimer.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit