Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dimethylnaphthidine 97.0+%, TCI America™
SDP

Catalog No. D07545G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 13138-48-2
Molecular Formula C22H20N2
Molecular Weight (g/mol) 312.42
MDL Number MFCD00042706
InChI Key FTEGMIZQLGXBNA-UHFFFAOYSA-N
PubChem CID 83166
IUPAC Name 3,3'-dimethyl-[1,1'-binaphthalene]-4,4'-diamine
SMILES CC1=CC(=C2C=CC=CC2=C1N)C1=C2C=CC=CC2=C(N)C(C)=C1

Specifications

Melting Point 232°C
Quantity 5 g
Formula Weight 312.42
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dimethylnaphthidine [for Colorimetric Determination of Cl in Water]
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.