Alfa Aesar™ 3,3-Dimethyloxetane, 98%

Manufacturer: Alfa Aesar™ H27370AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH27370AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6921-35-3
    Molecular Formula C5H10O
    Formula Weight 86.13
    MDL Number MFCD00005168
    UN Number UN3271
    Chemical Name or Material 3,3-Dimethyloxetane

    Specifications

    Formula Weight 86.13
    UN Number UN3271
    Flash Point −9°C (16°F)
    Quantity 5mL
    Molecular Formula C5H10O
    MDL Number MFCD00005168
    CAS 6921-35-3
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    Boiling Point 81°C
    Chemical Name or Material 3,3-Dimethyloxetane
    Assay 98%

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit