Alfa Aesar™ 3,3-Dimethylpentane, 99%

Manufacturer: Alfa Aesar™ 4370606

Quantity 5g

 View more versions of this product

Catalog No. AA4370606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 562-49-2
    Molecular Formula C7H16
    Formula Weight 100.21
    MDL Number MFCD00009322
    UN Number UN1206
    Chemical Name or Material 3,3-Dimethylpentane

    Specifications

    Formula Weight 100.21
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Density 0.69g/mL
    Flash Point −6°C (21°F)
    Physical Form Liquid
    Molecular Formula C7H16
    MDL Number MFCD00009322
    Melting Point −135°C
    TSCA TSCA
    UN Number UN1206
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
    Boiling Point 86°C
    Quantity 5g
    CAS 562-49-2
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Chemical Name or Material 3,3-Dimethylpentane
    Assay 99%

    GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit