Alfa Aesar™ 3,3-Diphenyl-L-alanine, 95%

Manufacturer: Alfa Aesar™ H52169MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52169MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 149597-92-2
    Molecular Formula C15H15NO2
    Formula Weight 241.29
    MDL Number MFCD01631989
    UN Number UN2811
    Synonym (S)-3-(3-Chlorophenyl)-3,3-diphenylpropionic acid; H-3,3-Bip-OH
    Chemical Name or Material 3,3-Diphenyl-L-alanine

    Specifications

    Formula Weight 241.29
    Molecular Formula C15H15NO2
    MDL Number MFCD01631989
    CAS 149597-92-2
    UN Number UN2811
    Synonym (S)-3-(3-Chlorophenyl)-3,3-diphenylpropionic acid; H-3,3-Bip-OH
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Quantity 250mg
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3,3-Diphenyl-L-alanine
    Assay 95%

    Description

    Applications
    3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit