Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dithiodipropionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
1119-62-6
C6H10O4S2
210.26
MFCD00002780
YCLSOMLVSHPPFV-UHFFFAOYSA-N
3,3'-dithiodipropionic acid,dithiodipropionic acid,propanoic acid, 3,3'-dithiobis,3,3-dithiodipropionic acid,3,3'-dithiobis dipropionic acid,unii-42op5b2ni6,3,3'-dithiobispropionic acid,propionic acid, 3,3'-dithiodi,3,3'-disulfanediyldipropanoic acid,3-2-carboxyethyl disulfanyl propanoic acid
95116
3-(2-carboxyethyldisulfanyl)propanoic acid
C(CSSCCC(=O)O)C(=O)O
1119-62-6
C6H10O4S2
210.26
MFCD00002780
YCLSOMLVSHPPFV-UHFFFAOYSA-N
3,3'-dithiodipropionic acid,dithiodipropionic acid,propanoic acid, 3,3'-dithiobis,3,3-dithiodipropionic acid,3,3'-dithiobis dipropionic acid,unii-42op5b2ni6,3,3'-dithiobispropionic acid,propionic acid, 3,3'-dithiodi,3,3'-disulfanediyldipropanoic acid,3-2-carboxyethyl disulfanyl propanoic acid
95116
3-(2-carboxyethyldisulfanyl)propanoic acid
C(CSSCCC(=O)O)C(=O)O
Quantity 50g
Melting Point 155°C to 158°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 98
Assay Percent Range 98%
Linear Formula S2(CH2CH2CO2H)2
Beilstein 03, IV, 736
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,benzene
Formula Weight 210.26
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material 3, 3'-Dithiodipropionic acid, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-284-0

TSCA : TSCA