Learn More
3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals
Description
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.
Solubility
Soluble in chloroform and methanol.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Melting Point | 110°C to 114°C |
Quantity | 5 g |
Beilstein | 1947445 |
Solubility Information | Soluble in chloroform and methanol. |
Formula Weight | 166.18 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-(3-Hydroxyphenyl)propionic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 210-692-8 |
RTECSNumber | MW5340000 |
TSCA | No |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.