Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Methoxyphenyl)propionic acid, 99%, Acros Organics™

Click to view available options
Quantity:
5g
25g
10516-71-9
C10H12O3
180.21
BJJQJLOZWBZEGA-UHFFFAOYSA-N
3-3-methoxyphenyl propionic acid,3-3-methoxyphenyl propanoic acid,3-m-methoxyphenyl propionic acid,3-methoxybenzenepropanoic acid,benzenepropanoic acid, 3-methoxy,3-3-methoxy-phenyl-propionic acid,3-methoxy-benzenepropanoic acid,3-3-methoxyphenyl propanoate,m-methoxyhydrocinnamate,3-methoxydihydrocinnamate
66336
3-(3-methoxyphenyl)propanoic acid
COC1=CC=CC(=C1)CCC(=O)O
10516-71-9
C10H12O3
180.21
BJJQJLOZWBZEGA-UHFFFAOYSA-N
3-3-methoxyphenyl propionic acid,3-3-methoxyphenyl propanoic acid,3-m-methoxyphenyl propionic acid,3-methoxybenzenepropanoic acid,benzenepropanoic acid, 3-methoxy,3-3-methoxy-phenyl-propionic acid,3-methoxy-benzenepropanoic acid,3-3-methoxyphenyl propanoate,m-methoxyhydrocinnamate,3-methoxydihydrocinnamate
66336
3-(3-methoxyphenyl)propanoic acid
COC1=CC=CC(=C1)CCC(=O)O
Quantity 25g
Melting Point 43.0°C to 46.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 180.21
Percent Purity 99%
Chemical Name or Material 3-(3-Methoxyphenyl)propionic acid

EINECSNumber : 234-049-6