Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3-Pentamethylene glutarimide, ≥98%

Click to view available options
Quantity:
1g
5g
25g
1130-32-1
C10H15NO2
181.235
MFCD00023872
FNIPRNMPSXNBDI-UHFFFAOYSA-N
3,3-pentamethylene glutarimide,3-azaspiro 5.5 undecane-2,4-dione,1,1-cyclohexanediacetimide,3,3-pentamethyleneglutarimide,3-azaspiro 5,5 undecan-2,4-dione,3-azaspiro 5.5 undecane-2,4-dione,9-azaspiro 5.5 undecane-8,10-dione,beta,beta-pentamethylen-glutarsaeureimid german,5-21-10-00098 beilstein handbook reference,beta,beta-pentamethylen-glutarsaeureimid
14324
3-azaspiro[5.5]undecane-2,4-dione
C1CCC2(CC1)CC(=O)NC(=O)C2
1130-32-1
C10H15NO2
181.235
MFCD00023872
FNIPRNMPSXNBDI-UHFFFAOYSA-N
3,3-pentamethylene glutarimide,3-azaspiro 5.5 undecane-2,4-dione,1,1-cyclohexanediacetimide,3,3-pentamethyleneglutarimide,3-azaspiro 5,5 undecan-2,4-dione,3-azaspiro 5.5 undecane-2,4-dione,9-azaspiro 5.5 undecane-8,10-dione,beta,beta-pentamethylen-glutarsaeureimid german,5-21-10-00098 beilstein handbook reference,beta,beta-pentamethylen-glutarsaeureimid
14324
3-azaspiro[5.5]undecane-2,4-dione
C1CCC2(CC1)CC(=O)NC(=O)C2
Quantity 25g
Melting Point 168°C to 170°C
Beilstein 383782
Formula Weight 181.23
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material 3,3-Pentamethylene glutarimide

RTECSNumber : CM0215000

storageNote1 : -30° to -10°C