Alfa Aesar™ 3,3-Tetramethyleneglutarimide, 98%

Manufacturer: Alfa Aesar™ B2486406

Quantity 5g

 View more versions of this product

Catalog No. AAB2486406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1075-89-4
    Molecular Formula C9H13NO2
    Formula Weight 167.21
    MDL Number MFCD00023871
    UN Number UN2811
    Beilstein 383702
    Synonym 8-Azaspiro[4.5]decane-7,9-dione; 1,1-Cyclopentanediacetimide
    Chemical Name or Material 3,3-Tetramethyleneglutarimide

    Specifications

    Formula Weight 167.21
    Odor Odorless
    Quantity 5g
    Molecular Formula C9H13NO2
    MDL Number MFCD00023871
    CAS 1075-89-4
    UN Number UN2811
    Beilstein 383702
    Synonym 8-Azaspiro[4.5]decane-7,9-dione; 1,1-Cyclopentanediacetimide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P260-P280f-P273-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Melting Point 153° to 156°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3,3-Tetramethyleneglutarimide
    Assay 98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P260-P280f-P273-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit