Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3-Tetramethyleneglutarimide, 98%

Click to view available options
Quantity:
5g
25g
1075-89-4
C9H13NO2
167.208
MFCD00023871
YRTHJMQKDCXPAY-UHFFFAOYSA-N
3,3-tetramethyleneglutarimide,8-azaspiro 4.5 decane-7,9-dione,azaspirodecanedione,1,1-cyclopentanediacetimide,8-aza-spiro 4.5 decane-7,9-dione,buspirone impurity k,dsstox_cid_31405,dsstox_rid_97291,dsstox_gsid_57616
136843
8-azaspiro[4.5]decane-7,9-dione
C1CCC2(C1)CC(=O)NC(=O)C2
1075-89-4
C9H13NO2
167.208
MFCD00023871
YRTHJMQKDCXPAY-UHFFFAOYSA-N
3,3-tetramethyleneglutarimide,8-azaspiro 4.5 decane-7,9-dione,azaspirodecanedione,1,1-cyclopentanediacetimide,8-aza-spiro 4.5 decane-7,9-dione,buspirone impurity k,dsstox_cid_31405,dsstox_rid_97291,dsstox_gsid_57616
136843
8-azaspiro[4.5]decane-7,9-dione
C1CCC2(C1)CC(=O)NC(=O)C2
Quantity 25g
Odor Odorless
Melting Point 153°C to 156°C
UN Number UN2811
Beilstein 383702
Formula Weight 167.21
Percent Purity 98%
Chemical Name or Material 3,3-Tetramethyleneglutarimide

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P260-P280f-P273-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.