Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3'-Thiodipropionic acid, 98%

Click to view available options
Quantity:
250g
1000g
111-17-1
C6H10O4S
178.202
MFCD00002781
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3,3'-thiodipropionic acid,thiodipropionic acid,propanoic acid, 3,3'-thiobis,bis 2-carboxyethyl sulfide,tyox a,thiodihydracrylic acid,4-thiaheptanedioic acid,tdpa,sulfide, bis 2-carboxyethyl,3,3'-thiodipropanoic acid
8096
3-(2-carboxyethylsulfanyl)propanoic acid
C(CSCCC(=O)O)C(=O)O
111-17-1
C6H10O4S
178.202
MFCD00002781
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3,3'-thiodipropionic acid,thiodipropionic acid,propanoic acid, 3,3'-thiobis,bis 2-carboxyethyl sulfide,tyox a,thiodihydracrylic acid,4-thiaheptanedioic acid,tdpa,sulfide, bis 2-carboxyethyl,3,3'-thiodipropanoic acid
8096
3-(2-carboxyethylsulfanyl)propanoic acid
C(CSCCC(=O)O)C(=O)O
Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

Solubility
Soluble in acetone, ethanol. Slightly soluble in water.

Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
Melting Point 127°C to 134°C
Quantity 250g
Linear Formula S(CH2CH2CO2H)2
Beilstein 1210299
Merck Index 14,9332
Solubility Information Soluble in acetone,ethanol. Slightly soluble in water.
Formula Weight 178.21
Percent Purity 98%
Chemical Name or Material 3,3'-Thiodipropionic acid

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-841-3

RTECSNumber : UF7990000

TSCA : Yes

Recommended Storage : Ambient temperatures