Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(Trifluoromethyl)phenoxy]benzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
2.5g
5g
Packaging:
Glass bottle
78725-46-9
C14H9F3O2
266.219
MFCD00003356
RPORRWJOHNQOHN-UHFFFAOYSA-N
3-3-trifluoromethyl phenoxy benzaldehyde,3-3-trifluoromethylphenoxy benzaldehyde,3-3'-trifluoromethyl phenoxy benzaldehyde,benzaldehyde,3-3-trifluoromethyl phenoxy,benzaldehyde, 3-3-trifluoromethyl phenoxy,acmc-1bgal,rporrwjohnqohn-uhfffaoysa,m-m-trifluoromethyl phenoxy benzaldehyde
522711
3-[3-(trifluoromethyl)phenoxy]benzaldehyde
C1=CC(=CC(=C1)OC2=CC=CC(=C2)C(F)(F)F)C=O
78725-46-9
C14H9F3O2
266.219
MFCD00003356
RPORRWJOHNQOHN-UHFFFAOYSA-N
3-3-trifluoromethyl phenoxy benzaldehyde,3-3-trifluoromethylphenoxy benzaldehyde,3-3'-trifluoromethyl phenoxy benzaldehyde,benzaldehyde,3-3-trifluoromethyl phenoxy,benzaldehyde, 3-3-trifluoromethyl phenoxy,acmc-1bgal,rporrwjohnqohn-uhfffaoysa,m-m-trifluoromethyl phenoxy benzaldehyde
522711
3-[3-(trifluoromethyl)phenoxy]benzaldehyde
C1=CC(=CC(=C1)OC2=CC=CC(=C2)C(F)(F)F)C=O
Color Yellow
Density 1.2800g/mL
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5365 to 1.5385 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CF3C6H4OC6H4CHO
Formula Weight 266.21
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-[3-(Trifluoromethyl)phenoxy]benzaldehyde, 97%
Show More Show Less