Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(Trifluoromethyl)phenyl]propanol, 98%, Alfa Aesar™

Click to view available options
Quantity:
250mg
78573-45-2
C10H11F3O
204.192
MFCD08706408
QWXKQVIMGVVIBX-UHFFFAOYSA-N
3-3-trifluoromethyl phenyl propan-1-ol,3-3-trifluoromethyl phenyl propan-1-ol,3-3'-trifluoromethyl phenyl propanol,3-3-trifluoromethyl phenyl-1-propanol,3-trifluoromethyl benzenepropanol,3-3-trifluoromethyl-phenyl-propan-1-ol,3-3'-trifluoromethylphenyl propanol,benzenepropanol, 3-trifluoromethyl,pubchem13744,benzenepropanol,3-trifluoromethyl
18755618
CHEBI:48528
3-[3-(trifluoromethyl)phenyl]propan-1-ol
C1=CC(=CC(=C1)C(F)(F)F)CCCO
78573-45-2
C10H11F3O
204.192
MFCD08706408
QWXKQVIMGVVIBX-UHFFFAOYSA-N
3-3-trifluoromethyl phenyl propan-1-ol,3-3-trifluoromethyl phenyl propan-1-ol,3-3'-trifluoromethyl phenyl propanol,3-3-trifluoromethyl phenyl-1-propanol,3-trifluoromethyl benzenepropanol,3-3-trifluoromethyl-phenyl-propan-1-ol,3-3'-trifluoromethylphenyl propanol,benzenepropanol, 3-trifluoromethyl,pubchem13744,benzenepropanol,3-trifluoromethyl
18755618
CHEBI:48528
3-[3-(trifluoromethyl)phenyl]propan-1-ol
C1=CC(=CC(=C1)C(F)(F)F)CCCO
Quantity 250mg
Color Colorless
Boiling Point 68°C to 71°C (0.4mmHg)
Density 1.10g/mL
Formula Weight 204.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol