Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
210907-84-9
C12H18BNO2
219.09
MFCD03453668
YMXIIVIQLHYKOT-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-aminophenylboronic acid pinacol ester,benzenamine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-aminophenylboronic acid, pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-aminobenzeneboronic acid pinacol ester,3-aminophenylboronic acid, pinacol cyclic ester,3-aminophenylboronicacidpinacolester,apbe,amtb060
2734655
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N
210907-84-9
C12H18BNO2
219.09
MFCD03453668
YMXIIVIQLHYKOT-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-aminophenylboronic acid pinacol ester,benzenamine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-aminophenylboronic acid, pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-aminobenzeneboronic acid pinacol ester,3-aminophenylboronic acid, pinacol cyclic ester,3-aminophenylboronicacidpinacolester,apbe,amtb060
2734655
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N
Melting Point 89.0°C to 94.0°C
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble in water.
Formula Weight 219.09
Percent Purity 97%
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C