Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
329214-79-1
C11H16BNO2
205.06
MFCD03084758
XEMDFESAXKSEGI-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,3-pyridineboronic acid pinacol ester,pyridine-3-boronic acid pinacol ester,3-pyridylboronic acid pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pyridine-3-boronic acid, pinacol ester,2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine,pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
2734656
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN=CC=C2
329214-79-1
C11H16BNO2
205.06
MFCD03084758
XEMDFESAXKSEGI-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,3-pyridineboronic acid pinacol ester,pyridine-3-boronic acid pinacol ester,3-pyridylboronic acid pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pyridine-3-boronic acid, pinacol ester,2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine,pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
2734656
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN=CC=C2
Quantity 25g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Melting Point 103°C to 108°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble
Formula Weight 205.06
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.