Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trichloroaniline, 97%

Click to view available options
Quantity:
1g
5g
25g
634-91-3
C6H4Cl3N
196.455
MFCD00007769
XOGYQVITULCUGU-UHFFFAOYSA-N
benzenamine, 3,4,5-trichloro,ccris 2398,aniline, 3,4,5-trichloro,3,4,5 trichloroaniline,acmc-1bag0,3,4,5-trichloro-aniline,3,4,5-trichlorophenylamine,3,4,5-trichlorobenzenamine,3,4,5-tris chloranyl aniline,3,4,5-trichloroaniline
12469
3,4,5-trichloroaniline
C1=C(C=C(C(=C1Cl)Cl)Cl)N
634-91-3
C6H4Cl3N
196.455
MFCD00007769
XOGYQVITULCUGU-UHFFFAOYSA-N
benzenamine, 3,4,5-trichloro,ccris 2398,aniline, 3,4,5-trichloro,3,4,5 trichloroaniline,acmc-1bag0,3,4,5-trichloro-aniline,3,4,5-trichlorophenylamine,3,4,5-trichlorobenzenamine,3,4,5-tris chloranyl aniline,3,4,5-trichloroaniline
12469
3,4,5-trichloroaniline
C1=C(C=C(C(=C1Cl)Cl)Cl)N
Applications
3,4,5-Trichloroaniline was used in the determination of substituted aromatic amines in sediment or water samples by capillary column gas chromatography.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Melting Point 98°C to 100°C
Quantity 25g
UN Number UN2018
Formula Weight 196.46
Percent Purity 97%
Chemical Name or Material 3,4,5-Trichloroaniline

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID