Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trichloropyridine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
33216-52-3
C5H2Cl3N
182.44
KKWRVUBDCJQHBZ-UHFFFAOYSA-N
pyridine, 3,4,5-trichloro,3,4,5-trichloro-pyridine,pubchem11021,acmc-209hzc,ksc497k5t,3,4,5-trichloropyridine,3,4,5-trichloro pyridine,3,4,5-trichloropyridine 1g
3786531
3,4,5-trichloropyridine
C1=C(C(=C(C=N1)Cl)Cl)Cl
33216-52-3
C5H2Cl3N
182.44
KKWRVUBDCJQHBZ-UHFFFAOYSA-N
pyridine, 3,4,5-trichloro,3,4,5-trichloro-pyridine,pubchem11021,acmc-209hzc,ksc497k5t,3,4,5-trichloropyridine,3,4,5-trichloro pyridine,3,4,5-trichloropyridine 1g
3786531
3,4,5-trichloropyridine
C1=C(C(=C(C=N1)Cl)Cl)Cl
Packaging Glass Bottle
Melting Point 72.0°C to 73.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 182.44
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridine