Alfa Aesar™ 3,4,5-Trifluoroanisole, 97%

Manufacturer: Alfa Aesar™ H2642003

Quantity 1g

 View more versions of this product

Catalog No. AAH2642003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 203245-17-4
    Molecular Formula C7H5F3O
    Formula Weight 162.11
    MDL Number MFCD00042397
    UN Number UN3271
    Synonym 5-Methoxy-1,2,3-trifluorobenzene
    Chemical Name or Material 3,4,5-Trifluoroanisole

    Specifications

    Formula Weight 162.11
    UN Number UN3271
    Flash Point 52°C (126°F)
    Melting Point ≈20°C
    Molecular Formula C7H5F3O
    MDL Number MFCD00042397
    Synonym 5-Methoxy-1,2,3-trifluorobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    Boiling Point 39°C (1mmHg)
    Quantity 1g
    CAS 203245-17-4
    Chemical Name or Material 3,4,5-Trifluoroanisole
    Assay 97%

    Description

    Applications
    3,4,5-Trifluoroanisole is used as a pharmaceutical intermediate.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit