Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluoroanisole, 97%

Manufacturer:  Alfa Aesar™ H2642006

Quantity 5g

 View more versions of this product

Catalog No. AAH2642006


  Check Availability
Add to cart

Specifications

Description

Applications
3,4,5-Trifluoroanisole is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

203245-17-4
162.11
UN3271
3,4,5-Trifluoroanisole
C7H5F3O
MFCD00042397
5-Methoxy-1,2,3-trifluorobenzene

Specifications

52°C (126°F)
≈20°C
203245-17-4
162.11
UN3271
3,4,5-Trifluoroanisole
39°C (1mmHg)
5g
C7H5F3O
MFCD00042397
5-Methoxy-1,2,3-trifluorobenzene
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit