Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorobenzaldehyde, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
132123-54-7
C7H3F3O
160.09
MFCD00083523
NLAVHUUABUFSIG-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trifluoro,3,4,5-trifluoro-benzaldehyde,3,4,5-ttrifluorobenzaldehyde,acmc-209bon,intermediates-zcf02014,3,4,5-triflourobenzaldehyde,3,4,5-trifluorobrnzaldehyde,ksc182i0f,3,4,5-trifluorobenzaldehyde,benzaldehyde, 3,4,5-trifluoro-9ci
2776954
3,4,5-trifluorobenzaldehyde
C1=C(C=C(C(=C1F)F)F)C=O
132123-54-7
C7H3F3O
160.09
MFCD00083523
NLAVHUUABUFSIG-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trifluoro,3,4,5-trifluoro-benzaldehyde,3,4,5-ttrifluorobenzaldehyde,acmc-209bon,intermediates-zcf02014,3,4,5-triflourobenzaldehyde,3,4,5-trifluorobrnzaldehyde,ksc182i0f,3,4,5-trifluorobenzaldehyde,benzaldehyde, 3,4,5-trifluoro-9ci
2776954
3,4,5-trifluorobenzaldehyde
C1=C(C=C(C(=C1F)F)F)C=O
Specific Gravity 1.42
Quantity 1g
Boiling Point 174.0°C
Melting Point 25.3°C
Color Colorless to Yellow
Packaging Glass bottle
Flash Point 63°C
Density 1.4200g/mL
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula F3C6H2CHO
Solubility Information Solubility in water: 2.2g/L (20°C)
Formula Weight 160.09
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning