Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluorobenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
134227-45-5
C7H2F3N
157.095
MFCD00074948
XFKYJMGXZXJYBS-UHFFFAOYSA-N
3,4,5-trifluoro-1-cyano benzene,benzonitrile, 3,4,5-trifluoro,3,4,5-trifluoro-benzonitrile,3,4,5-trifluorobenzenecarbonitrile,pubchem1573,intermediates-zcf02038,acmc-1bz57,ksc493s4j,3,4,5-?trifluorobenzonitrile,3,4,5-trifluorobenzonitrile
593814
3,4,5-trifluorobenzonitrile
C1=C(C=C(C(=C1F)F)F)C#N
134227-45-5
C7H2F3N
157.095
MFCD00074948
XFKYJMGXZXJYBS-UHFFFAOYSA-N
3,4,5-trifluoro-1-cyano benzene,benzonitrile, 3,4,5-trifluoro,3,4,5-trifluoro-benzonitrile,3,4,5-trifluorobenzenecarbonitrile,pubchem1573,intermediates-zcf02038,acmc-1bz57,ksc493s4j,3,4,5-?trifluorobenzonitrile,3,4,5-trifluorobenzonitrile
593814
3,4,5-trifluorobenzonitrile
C1=C(C=C(C(=C1F)F)F)C#N
Quantity 1g
Melting Point 44°C to 47°C
Flash Point 62°C (143°F)
Boiling Point 72°C to 73°C (10mmHg)
UN Number UN3439
Beilstein 7702244
Formula Weight 157.1
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.