Alfa Aesar™ 3,4,5-Trifluorobenzyl alcohol, 97%

Manufacturer: Alfa Aesar™ B2461706

Quantity 5g

 View more versions of this product

Catalog No. AAB2461706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 220227-37-2
    Molecular Formula C7H5F3O
    Formula Weight 162.11
    MDL Number MFCD00083527
    Beilstein 8681069
    Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol

    Specifications

    Formula Weight 162.11
    Molecular Formula C7H5F3O
    CAS 220227-37-2
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 98°C (208°F)
    Boiling Point 203° to 204°C
    Quantity 5g
    MDL Number MFCD00083527
    Beilstein 8681069
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol
    Assay 97%

    Description

    Applications
    3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.

    Solubility
    Soluble in water 100 g/L.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit