Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluorobenzyl alcohol, 97%

Click to view available options
Quantity:
1g
5g
220227-37-2
C7H5F3O
162.111
MFCD00083527
HRSFRSLKOPFWMZ-UHFFFAOYSA-N
3,4,5-trifluorobenzyl alcohol,3,4,5-trifluorophenyl methanol,3,4,5-trifluorobenzenemethanol,benzenemethanol, 3,4,5-trifluoro,3,4,5-trifluorophenyl methan-1-ol,3,4,5-trifluoro benzenemethanol,acmc-20angf,3,4,5-trifluorobenzylalcohol,3,4,5-trifluoro-benzenemethanol,benzenemethanol,3,4,5-trifluoro
2777040
(3,4,5-trifluorophenyl)methanol
C1=C(C=C(C(=C1F)F)F)CO
220227-37-2
C7H5F3O
162.111
MFCD00083527
HRSFRSLKOPFWMZ-UHFFFAOYSA-N
3,4,5-trifluorobenzyl alcohol,3,4,5-trifluorophenyl methanol,3,4,5-trifluorobenzenemethanol,benzenemethanol, 3,4,5-trifluoro,3,4,5-trifluorophenyl methan-1-ol,3,4,5-trifluoro benzenemethanol,acmc-20angf,3,4,5-trifluorobenzylalcohol,3,4,5-trifluoro-benzenemethanol,benzenemethanol,3,4,5-trifluoro
2777040
(3,4,5-trifluorophenyl)methanol
C1=C(C=C(C(=C1F)F)F)CO
Applications
3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.

Solubility
Soluble in water 100 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 1g
Flash Point 98°C (208°F)
Boiling Point 203°C to 204°C
Beilstein 8681069
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning