Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluorobenzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
220141-72-0
C7H4BrF3
225.008
MFCD00083528
MPEKZIYOLQCWLL-UHFFFAOYSA-N
3,4,5-trifluorobenzyl bromide,5-bromomethyl-1,2,3-trifluorobenzene,3,4,5-trifluorobenzylbromide,alpha-bromo-3,4,5-trifluorotoluene,3,4,5-trifluoro benzyl bromide,benzene, 5-bromomethyl-1,2,3-trifluoro,a-bromo-3,4,5-trifluorotoluene,pubchem4670,acmc-1cupe
2777062
5-(bromomethyl)-1,2,3-trifluorobenzene
C1=C(C=C(C(=C1F)F)F)CBr
220141-72-0
C7H4BrF3
225.008
MFCD00083528
MPEKZIYOLQCWLL-UHFFFAOYSA-N
3,4,5-trifluorobenzyl bromide,5-bromomethyl-1,2,3-trifluorobenzene,3,4,5-trifluorobenzylbromide,alpha-bromo-3,4,5-trifluorotoluene,3,4,5-trifluoro benzyl bromide,benzene, 5-bromomethyl-1,2,3-trifluoro,a-bromo-3,4,5-trifluorotoluene,pubchem4670,acmc-1cupe
2777062
5-(bromomethyl)-1,2,3-trifluorobenzene
C1=C(C=C(C(=C1F)F)F)CBr
Boiling Point 44°C to 46°C (1mmHg)
Flash Point 65°C (149°F)
Quantity 5g
UN Number UN3265
Beilstein 8680985
Formula Weight 225.01
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzyl bromide

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.