Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluorocinnamic acid, 97+%

Click to view available options
Quantity:
1g
5g
152152-19-7
C9H5F3O2
202.132
MFCD00083529
PHIWFMZBVZFEQJ-OWOJBTEDSA-N
3,4,5-trifluorocinnamic acid,2e-3-3,4,5-trifluorophenyl prop-2-enoic acid,3-3,4,5-trifluoro-phenyl-acrylic acid,3,4,5-trifluorocinnamicacid,pubchem4351,rarechem bk hw 0056,trans-3,4,5-trifluorocinnamic acid,e-3-3,4,5-trifluorophenyl acrylic acid,e-3-3,4,5-trifluorophenyl prop-2-enoic acid,2-propenoic acid, 3-3,4,5-trifluorophenyl-, 2e
5708779
(E)-3-(3,4,5-trifluorophenyl)prop-2-enoic acid
C1=C(C=C(C(=C1F)F)F)C=CC(=O)O
152152-19-7
C9H5F3O2
202.132
MFCD00083529
PHIWFMZBVZFEQJ-OWOJBTEDSA-N
3,4,5-trifluorocinnamic acid,2e-3-3,4,5-trifluorophenyl prop-2-enoic acid,3-3,4,5-trifluoro-phenyl-acrylic acid,3,4,5-trifluorocinnamicacid,pubchem4351,rarechem bk hw 0056,trans-3,4,5-trifluorocinnamic acid,e-3-3,4,5-trifluorophenyl acrylic acid,e-3-3,4,5-trifluorophenyl prop-2-enoic acid,2-propenoic acid, 3-3,4,5-trifluorophenyl-, 2e
5708779
(E)-3-(3,4,5-trifluorophenyl)prop-2-enoic acid
C1=C(C=C(C(=C1F)F)F)C=CC(=O)O
Quantity 1g
Melting Point 195°C to 199°C
Formula Weight 202.13
Percent Purity ≥97%
Chemical Name or Material 3,4,5-Trifluorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning