Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trifluorophenol, 97%

Click to view available options
Quantity:
1g
5g
99627-05-1
C6H3F3O
148.084
MFCD00042427
ZRTWIJKGTUGZJY-UHFFFAOYSA-N
phenol, 3,4,5-trifluoro,3,4,5-trifluoro-phenol,pubchem2316,3,4,5-trifluoropheno,acmc-209sdi,ksc486m6r,3 , 4 , 5-trifluorophenol,phenol, 3,4,5-trifluoro-9ci,3,4,5-trifluorophenol
2777943
3,4,5-trifluorophenol
C1=C(C=C(C(=C1F)F)F)O
99627-05-1
C6H3F3O
148.084
MFCD00042427
ZRTWIJKGTUGZJY-UHFFFAOYSA-N
phenol, 3,4,5-trifluoro,3,4,5-trifluoro-phenol,pubchem2316,3,4,5-trifluoropheno,acmc-209sdi,ksc486m6r,3 , 4 , 5-trifluorophenol,phenol, 3,4,5-trifluoro-9ci,3,4,5-trifluorophenol
2777943
3,4,5-trifluorophenol
C1=C(C=C(C(=C1F)F)F)O
Melting Point 52°C to 56°C
Boiling Point 177°C
Quantity 5g
UN Number UN3261
Beilstein 8830884
Formula Weight 148.09
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorophenol

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.